Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LYRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128301.2 | NP_001121773.1 | 3 | 554 | ||
NM_001128302.2 | NP_001121774.1 | 2 | 322 | |||
NM_001302835.1 | NP_001289764.1 | 4 | 498 | |||
NM_001302836.1 | NP_001289765.1 | 4 | 650 | |||
NM_020424.4 | NP_065157.1 | 3 | 473 | |||
XM_005255444.1 | XP_005255501.1 | 3 | 375 | |||
XM_011545900.2 | XP_011544202.1 | 2 | 122 | |||
XM_011545901.2 | XP_011544203.1 | 2 | 188 | |||
XM_011545902.1 | XP_011544204.1 | 3 | 459 | |||
XM_011545908.2 | XP_011544210.1 | 2 | 190 | |||
XM_017023475.1 | XP_016878964.1 | 3 | 875 | |||
XM_017023476.1 | XP_016878965.1 | 3 | 881 | |||
XM_017023478.1 | XP_016878967.1 | 2 | 180 | |||
XM_017023479.1 | XP_016878968.1 | 3 | 632 | |||
XM_017023480.1 | XP_016878969.1 | 5 | 718 | |||
XM_017023481.1 | XP_016878970.1 | 3 | 347 | |||
XM_017023482.1 | XP_016878971.1 | 2 | 401 | |||
GenBank mRNA | AK310055.1 | - | 2 | 223 | ||
AK311321.1 | - | 3 | 375 | |||
AK311881.1 | - | 2 | 189 | |||
AV714321.1 | - | 2 | 168 | |||
BC017039.1 | - | 4 | 498 | 3849248 | ||
BI547471.1 | - | 2 | 240 | |||
BI825882.1 | - | 4 | 497 | |||
CD237780.1 | - | 2 | 192 | |||
CX163936.1 | - | 2 | 199 | |||
DA572182.1 | - | 3 | 439 |