Search
Search

ARID1B| Gene Symbol: ARID1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017519.2 | NP_059989.2 | 4 | 1966 | ||
| NM_020732.3 | NP_065783.3 | 5 | 2005 | |||
| XM_005267069.4 | XP_005267126.2 | 4 | 2041 | |||
| XM_011535984.2 | XP_011534286.2 | 5 | 2081 | |||
| XM_017011103.1 | XP_016866592.1 | 5 | 2081 | |||
| XM_017011104.1 | XP_016866593.1 | 5 | 2080 | |||
| XM_017011105.1 | XP_016866594.1 | 5 | 2081 | |||
| XM_017011106.1 | XP_016866595.1 | 5 | 2080 | |||
| XM_017011107.1 | XP_016866596.1 | 5 | 2081 | |||
| XM_017011108.1 | XP_016866597.1 | 5 | 2088 | |||
| GenBank mRNA | AF253515.1 | - | 4 | 1160 | ||
| AF468300.1 | - | 5 | 535 | |||
| AF521671.1 | - | 4 | 1753 | |||
| AK025945.1 | - | 2 | 232 | |||
| AK074180.1 | - | 1 | 74 | |||