Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DHX37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032656.3 | NP_116045.2 | 22 | 3013 | ||
XM_005253590.3 | XP_005253647.1 | 22 | 3017 | |||
GenBank mRNA | AB040950.1 | - | 19 | 2385 | ||
AK123968.1 | - | 2 | 1083 | |||
BC002575.2 | - | 19 | 2396 | |||
BC037964.1 | - | 22 | 2995 | 5736582 |