Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DENND1A SIRNA Assays ›
          Gene Symbol:
          DENND1A
          Gene Name:
          DENN domain containing 1A
          Gene Aliases:
          FAM31A, KIAA1608
          Chromosome Location:
          Chr.9: 123379654 - 123930158 on Build GRCh38
          UniGene ID:
          Hs.744995
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s33639

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DENND1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_020946.1 NP_065997.1 8, 9 733
          NM_024820.2 NP_079096.2 8, 9 733
          XM_005252109.2 XP_005252166.1 8, 9 582
          XM_005252111.4 XP_005252168.1 8, 9 1549
          XM_005252113.4 XP_005252170.1 8, 9 1538
          XM_006717195.3 XP_006717258.1 8, 9 1550
          XM_011518882.2 XP_011517184.1 8, 9 1552
          XM_011518883.2 XP_011517185.1 8, 9 1553
          XM_011518884.2 XP_011517186.1 7, 8 1503
          XM_011518885.2 XP_011517187.1 7, 8 1245
          XM_011518886.2 XP_011517188.1 8, 9 1550
          XM_011518887.2 XP_011517189.1 8, 9 1541
          XM_017014947.1 XP_016870436.1 7, 8 1508
          XM_017014948.1 XP_016870437.1 8, 9 582
          XM_017014949.1 XP_016870438.1 8, 9 1553
          XM_017014950.1 XP_016870439.1 8, 9 1552
          XM_017014951.1 XP_016870440.1 7, 8 526
          XM_017014952.1 XP_016870441.1 7, 8 709
          GenBank mRNA AB046828.2 - 6, 7 597
          AK024782.1 - 8, 9 721
          AK074151.1 - 3, 4 161
          AK225104.1 - 8, 9 721
          AK295710.1 - 6, 7 594
          BC009616.2 - 7, 8 701
          BC028061.1 - 7, 8 523
          BC039703.1 - 8, 9 586 4999484

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001191747, XM_008761757, XM_008761756.
          This siRNA also targets the following transcripts in Mouse: NM_146122, BC023016, AK136192, DQ448594, AK137530, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          protein transport
          endocytic recycling
          regulation of Rab protein signal transduction
          positive regulation of GTPase activity
          synaptic vesicle endocytosis
          Rab guanyl-nucleotide exchange factor activity
          SH3 domain binding
          phosphatidylinositol-3-phosphate binding
          phosphatidylinositol phosphate binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0