Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OSTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267818.1 | NP_001254747.1 | 3 | 391 | ||
NM_021227.3 | NP_067050.1 | 3 | 391 | |||
GenBank mRNA | AF068297.1 | - | 3 | 354 | ||
AF161425.1 | - | 3 | 375 | |||
AF201937.1 | - | 3 | 382 | |||
AF343342.1 | - | 3 | 320 | |||
AK312185.1 | - | 3 | 335 | |||
BC016321.1 | - | 3 | 379 | 4075646 | ||
BC054857.1 | - | 3 | 381 | 4093030 | ||
BM471860.1 | - | 319 | ||||
BM541430.1 | - | 3 | 342 | |||
BM546028.1 | - | 3 | 381 | |||
BQ435510.1 | - | 391 | ||||
CN430159.1 | - | 3 | 329 |