Search Thermo Fisher Scientific
Gene Symbol: MID1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098790.1 | NP_001092260.1 | 3 | 1589 | ||
NM_001098791.1 | NP_001092261.1 | 2 | 1485 | |||
NM_021242.5 | NP_067065.1 | 3 | 3364 | |||
GenBank mRNA | AF050158.1 | - | 1 | 395 | ||
AJ272057.1 | - | 2 | 1624 | |||
AK001248.1 | - | 2 | 1569 | |||
BC008908.2 | - | 2 | 2022 | 3638510 | ||
BC019332.1 | - | 3 | 1567 | 3541914 |