Search Thermo Fisher Scientific
Gene Symbol: SNX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021249.3 | NP_067072.3 | 6 | 740 | ||
NM_152233.2 | NP_689419.2 | 7 | 635 | |||
XM_006720224.1 | XP_006720287.1 | 6 | 522 | |||
GenBank mRNA | AF109364.1 | - | 2 | 118 | ||
AF121856.1 | - | 7 | 626 | |||
AK021456.1 | - | 2 | 122 | |||
AK292250.1 | - | 7 | 618 | |||
AK296213.1 | - | 8 | 763 | |||
AK308464.1 | - | 8 | 763 | |||
BC001798.1 | - | 6 | 740 | 3354467 | ||
BG722853.1 | - | 7 | 662 | |||
U83194.1 | - | 7 | 603 |