Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMBIM1 SIRNA Assays ›
          Gene Symbol:
          TMBIM1
          Gene Name:
          transmembrane BAX inhibitor motif containing 1
          Gene Aliases:
          LFG3, MST100, MSTP100, PP1201, RECS1
          Chromosome Location:
          Chr.2: 218274192 - 218292577 on Build GRCh38
          UniGene ID:
          Hs.591605
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s34463

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMBIM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321427.1 NP_001308356.1 6, 7 1159
          NM_001321428.1 NP_001308357.1 5, 6 570
          NM_001321429.1 NP_001308358.1 6, 7 746
          NM_001321430.1 NP_001308359.1 6, 7 686
          NM_001321432.1 NP_001308361.1 5, 6 510
          NM_001321433.1 NP_001308362.1 6, 7 683
          NM_001321435.1 NP_001308364.1 6, 7 757
          NM_001321436.1 NP_001308365.1 6, 7 761
          NM_001321438.1 NP_001308367.1 4, 5 520
          NM_022152.5 NP_071435.2 5, 6 585
          NR_135643.1 4, 5 343
          XM_011511627.1 XP_011509929.1 5, 6 938
          GenBank mRNA AB257295.1 - 5, 6 646
          AF193045.1 - 5, 6 499
          AK027755.1 - 3, 4 233
          AK075465.1 - 1, 2 60
          AK090618.1 - 6, 7 1157
          AK093911.1 - 3, 4 1097
          AK130380.1 - 5, 6 543
          AK130713.1 - 5, 6 521
          AK298123.1 - 6, 7 357
          AK298500.1 - 4, 5 279
          AK300597.1 - 4, 5 447
          BC007980.2 - 5, 6 499 3689664
          BC013428.1 - 5, 6 563 3899632
          BC026348.1 - 5, 6 520 4792398
          BC026693.2 - 5, 6 531
          CF994144.1 - 5, 6 547

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of catalytic activity
          negative regulation of establishment of protein localization to plasma membrane
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of Fas signaling pathway
          positive regulation of blood vessel remodeling
          death receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0