Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SOX17 SIRNA Assays ›
Gene Symbol:
SOX17
Gene Name:
SRY-box 17
Gene Aliases:
VUR3
Chromosome Location:
Chr.8: 54457935 - 54460896 on Build GRCh38
UniGene ID:
Hs.98367
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s34626
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SOX17
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022454.3
NP_071899.1
2
1291
GenBank mRNA
AB073988.1
-
2
1090
AK025905.1
-
2
1291
BC111770.1
-
2
479
Back To Top
More Information
Panther Classification:
Molecular Function -
HMG box transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
vasculogenesis
endoderm formation
inner cell mass cellular morphogenesis
heart looping
cardiogenic plate morphogenesis
embryonic heart tube morphogenesis
outflow tract morphogenesis
negative regulation of Wnt signaling pathway involved in heart development
regulation of transcription, DNA-templated
spermatogenesis
endodermal cell fate determination
rostrocaudal neural tube patterning
signal transduction involved in regulation of gene expression
negative regulation of cell growth
protein destabilization
embryonic heart tube development
cell migration involved in gastrulation
negative regulation of mesodermal cell fate specification
mRNA transcription from RNA polymerase II promoter
positive regulation of cell differentiation
positive regulation of protein catabolic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of embryonic development
embryonic foregut morphogenesis
positive regulation of skeletal muscle tissue development
stem cell fate specification
protein stabilization
canonical Wnt signaling pathway
endocardium formation
regulation of transcription from RNA polymerase II promoter involved in definitive endodermal cell fate specification
cardiac cell fate determination
heart formation
endocardial cell differentiation
common bile duct development
gall bladder development
endodermal digestive tract morphogenesis
renal system development
regulation of stem cell proliferation
negative regulation of canonical Wnt signaling pathway
regulation of stem cell division
regulation of cardiac cell fate specification
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription coactivator activity
protein binding
beta-catenin binding
transcription factor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-78lvj:80/100.66.75.111:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0