Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHID1 SIRNA Assays ›
          Gene Symbol:
          CHID1
          Gene Name:
          chitinase domain containing 1
          Gene Aliases:
          GL008, SI-CLP, SICLP
          Chromosome Location:
          Chr.11: 867859 - 915058 on Build GRCh38
          UniGene ID:
          Hs.144468
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s35346

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHID1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142674.1 NP_001136146.1 3, 4 364
          NM_001142675.1 NP_001136147.1 2, 3 474
          NM_001142677.1 NP_001136149.1 2, 3 260
          NM_023947.3 NP_076436.3 2, 3 260
          XM_005253072.2 XP_005253129.1 2, 3 347
          XM_005253073.4 XP_005253130.1 3, 4 348
          XM_006718282.2 XP_006718345.1 3, 4 335
          XM_011520299.1 XP_011518601.1 2, 3 571
          XM_011520300.1 XP_011518602.1 2, 3 571
          XM_011520301.2 XP_011518603.1 2, 3 571
          XM_011520302.2 XP_011518604.1 2, 3 571
          XM_017018166.1 XP_016873655.1 3, 4 421
          XM_017018167.1 XP_016873656.1 2, 3 226
          XM_017018168.1 XP_016873657.1 3, 4 347
          GenBank mRNA AF212229.1 - 3, 4 298
          AK027711.1 - 2, 3 209
          AK124697.1 - 2, 3 836
          AK225025.1 - 3, 4 300
          AK225398.1 - 3, 4 283
          AK297743.1 - 2, 3 341
          AK301245.1 - 2, 3 320
          AY037151.1 - 2, 3 221
          BC000001.2 - 3, 4 284 3504261
          BC095409.1 - 2, 3 196 30333427
          BI759317.1 - 2, 3 476
          BI818660.1 - 3, 4 364
          BI821281.1 - 2, 3 226
          DA983719.1 - 2, 3 431

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          chitin catabolic process
          innate immune response
          negative regulation of cytokine production involved in inflammatory response
          chitinase activity
          protein binding
          chitin binding
          oligosaccharide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-n8jtk:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0