Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C7orf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024067.3 | NP_076972.2 | 4 | 1120 | ||
XM_005249845.4 | XP_005249902.1 | 4 | 1138 | |||
XM_011515518.2 | XP_011513820.1 | 4 | 1138 | |||
XM_017012614.1 | XP_016868103.1 | 4 | 1666 | |||
XM_017012615.1 | XP_016868104.1 | 3 | 662 | |||
GenBank mRNA | AK056149.1 | - | 4 | 857 | ||
CA454040.1 | - | 1 | 277 |