Search
Search

| Gene Symbol: OR4F5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005484.1 | NP_001005484.1 | 1 | 224 | ||
| Gene Symbol: OR4F17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005240.1 | NP_001005240.1 | 1 | 224 | ||
| XM_005259648.1 | XP_005259705.1 | 2 | 367 | |||
| GenBank mRNA | AY972817.1 | - | 2 | 373 | ||
| AY972818.1 | - | 1 | 290 | |||
| BC136848.1 | - | 1 | 260 | 9020839 | ||
| BC136867.1 | - | 1 | 260 | 9020858 | ||
| Gene Symbol: OR4F4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004195.2 | NP_001004195.2 | 1 | 224 | ||
| GenBank mRNA | AY972819.1 | - | 1 | 290 | ||
| BC136907.1 | - | 1 | 260 | 9020898 | ||
| BC136908.1 | - | 1 | 260 | 9020899 | ||