Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CSF1 SIRNA Assays ›
          Gene Symbol:
          CSF1
          Gene Name:
          colony stimulating factor 1
          Gene Aliases:
          CSF-1, MCSF
          Chromosome Location:
          Chr.1: 109910506 - 109930994 on Build GRCh38
          UniGene ID:
          Hs.173894
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s3592

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CSF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000757.5 NP_000748.3 2 504
          NM_172210.2 NP_757349.1 2 504
          NM_172211.3 NP_757350.1 2 504
          NM_172212.2 NP_757351.1 2 504
          XM_017000369.1 XP_016855858.1 2 179
          GenBank mRNA AB208952.1 - 2 253
          AK291660.1 - 2 266
          BC021117.1 - 2 280 4586312
          DA414355.1 - 2 504
          DB287787.1 - 2 437
          M27087.1 - 2 215
          M37435.1 - 1 196
          M64592.1 - 2 91
          M76453.1 - 1 55
          U22386.1 - 2 135
          X05825.1 - 1 39

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          positive regulation of cell-matrix adhesion
          osteoclast proliferation
          developmental process involved in reproduction
          inflammatory response
          transmembrane receptor protein tyrosine kinase signaling pathway
          cell proliferation
          positive regulation of cell proliferation
          positive regulation of gene expression
          regulation of macrophage derived foam cell differentiation
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of macrophage chemotaxis
          hemopoiesis
          cell differentiation
          macrophage differentiation
          regulation of ossification
          osteoclast differentiation
          positive regulation of cell migration
          positive regulation of cellular protein metabolic process
          positive regulation of mononuclear cell proliferation
          macrophage colony-stimulating factor signaling pathway
          positive regulation of multicellular organism growth
          monocyte activation
          odontogenesis
          positive regulation of odontogenesis of dentin-containing tooth
          innate immune response
          positive regulation of macrophage differentiation
          positive regulation of monocyte differentiation
          positive regulation of osteoclast differentiation
          positive regulation of protein kinase activity
          positive regulation of Ras protein signal transduction
          homeostasis of number of cells within a tissue
          branching involved in mammary gland duct morphogenesis
          mammary gland fat development
          mammary duct terminal end bud growth
          positive regulation of macrophage colony-stimulating factor signaling pathway
          positive regulation of microglial cell migration
          cytokine activity
          macrophage colony-stimulating factor receptor binding
          protein binding
          growth factor activity
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0