Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AAGAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271885.1 | NP_001258814.1 | 2 | 83 | ||
NM_001271886.1 | NP_001258815.1 | 2 | 344 | |||
NM_024666.4 | NP_078942.3 | 2 | 199 | |||
XM_011522020.1 | XP_011520322.1 | 2 | 449 | |||
GenBank mRNA | AK021568.1 | - | 2 | 103 | ||
AK294402.1 | - | 2 | 80 | |||
AL136715.1 | - | 2 | 109 | |||
BC047026.1 | - | 2 | 122 | 5534878 | ||
BC058886.1 | - | 2 | 121 | 6503238 | ||
CR533540.1 | - | 2 | 94 | |||
DA160293.1 | - | 2 | 259 | |||
DA688370.1 | - | 2 | 199 | |||
DC315715.1 | - | 2 | 83 |