Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC7 SIRNA Assays ›
Gene Symbol:
CCDC7
Gene Name:
coiled-coil domain containing 7
Gene Aliases:
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location:
Chr.10: 32446082 - 32882864 on Build GRCh38
UniGene ID:
Hs.585464
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36231
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321115.1
NP_001308044.1
33
3796
XM_005252593.2
XP_005252650.1
14
1583
XM_006717505.1
XP_006717568.1
31
3291
XM_006717506.1
XP_006717569.1
30
3183
XM_011519663.1
XP_011517965.1
33
3504
XM_011519664.1
XP_011517966.1
33
3504
XM_011519666.1
XP_011517968.1
32
3420
XM_011519667.1
XP_011517969.1
33
3504
XM_011519668.1
XP_011517970.1
32
3414
XM_011519669.1
XP_011517971.1
32
3402
XM_011519670.1
XP_011517972.1
32
3396
XM_011519671.1
XP_011517973.1
32
3396
XM_011519672.1
XP_011517974.1
32
3393
XM_011519673.1
XP_011517975.1
32
3390
XM_011519674.1
XP_011517976.1
32
3396
XM_011519675.1
XP_011517977.1
31
3294
XM_011519676.1
XP_011517978.1
31
3282
XM_011519677.1
XP_011517979.1
32
3282
XM_011519678.1
XP_011517980.1
31
3279
XM_011519679.1
XP_011517981.1
30
2973
XM_011519680.2
XP_011517982.1
28
2639
XM_011519683.2
XP_011517985.1
22
2313
XM_011519684.2
XP_011517986.1
34
3585
XM_011519685.1
XP_011517987.1
25
2521
XM_011519687.1
XP_011517989.1
15
1428
XM_017016638.1
XP_016872127.1
33
3459
XM_017016639.1
XP_016872128.1
32
3393
XM_017016640.1
XP_016872129.1
31
3312
XM_017016641.1
XP_016872130.1
28
2933
XM_017016642.1
XP_016872131.1
28
3006
XM_017016643.1
XP_016872132.1
28
2623
XM_017016644.1
XP_016872133.1
33
3504
XM_017016645.1
XP_016872134.1
27
2547
XM_017016646.1
XP_016872135.1
24
2434
XM_017016647.1
XP_016872136.1
34
3588
XM_017016648.1
XP_016872137.1
20
2155
XM_017016649.1
XP_016872138.1
20
2095
XM_017016652.1
XP_016872141.1
17
1558
XM_017016653.1
XP_016872142.1
15
1468
GenBank mRNA
AK023093.1
-
13
1481
AK097668.1
-
6
537
AK301824.1
-
11
979
BC125091.1
-
11
1006
40118177
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0