Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCDC7 SIRNA Assays ›
          Gene Symbol:
          CCDC7
          Gene Name:
          coiled-coil domain containing 7
          Gene Aliases:
          BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
          Chromosome Location:
          Chr.10: 32446082 - 32882864 on Build GRCh38
          UniGene ID:
          Hs.585464
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s36233

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCDC7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321115.1 NP_001308044.1 32, 33 3786
          XM_005252593.2 XP_005252650.1 13, 14 1573
          XM_006717505.1 XP_006717568.1 30, 31 3281
          XM_006717506.1 XP_006717569.1 29, 30 3173
          XM_011519663.1 XP_011517965.1 32, 33 3494
          XM_011519664.1 XP_011517966.1 32, 33 3494
          XM_011519666.1 XP_011517968.1 31, 32 3410
          XM_011519667.1 XP_011517969.1 32, 33 3494
          XM_011519668.1 XP_011517970.1 31, 32 3404
          XM_011519669.1 XP_011517971.1 31, 32 3392
          XM_011519670.1 XP_011517972.1 31, 32 3386
          XM_011519671.1 XP_011517973.1 31, 32 3386
          XM_011519672.1 XP_011517974.1 31, 32 3383
          XM_011519673.1 XP_011517975.1 31, 32 3380
          XM_011519674.1 XP_011517976.1 31, 32 3386
          XM_011519675.1 XP_011517977.1 30, 31 3284
          XM_011519676.1 XP_011517978.1 30, 31 3272
          XM_011519677.1 XP_011517979.1 31, 32 3272
          XM_011519678.1 XP_011517980.1 30, 31 3269
          XM_011519679.1 XP_011517981.1 29, 30 2963
          XM_011519680.2 XP_011517982.1 27, 28 2629
          XM_011519683.2 XP_011517985.1 21, 22 2303
          XM_011519685.1 XP_011517987.1 24, 25 2511
          XM_011519687.1 XP_011517989.1 14, 15 1418
          XM_017016638.1 XP_016872127.1 32, 33 3449
          XM_017016639.1 XP_016872128.1 31, 32 3383
          XM_017016640.1 XP_016872129.1 30, 31 3302
          XM_017016641.1 XP_016872130.1 27, 28 2923
          XM_017016642.1 XP_016872131.1 27, 28 2996
          XM_017016643.1 XP_016872132.1 27, 28 2613
          XM_017016644.1 XP_016872133.1 32, 33 3494
          XM_017016645.1 XP_016872134.1 26, 27 2537
          XM_017016646.1 XP_016872135.1 23, 24 2424
          XM_017016648.1 XP_016872137.1 19, 20 2145
          XM_017016649.1 XP_016872138.1 19, 20 2085
          XM_017016652.1 XP_016872141.1 16, 17 1548
          XM_017016653.1 XP_016872142.1 14, 15 1458
          GenBank mRNA AK023093.1 - 12, 13 1471
          AK097668.1 - 5, 6 527
          AK301824.1 - 10, 11 969
          BC125091.1 - 10, 11 996 40118177

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-rvwhl:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0