Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CLMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024769.3 | NP_079045.1 | 2 | 526 | ||
XM_011542998.2 | XP_011541300.1 | 2 | 494 | |||
GenBank mRNA | AK026068.1 | - | 2 | 502 | ||
AK304843.1 | - | 2 | 473 | |||
AY326422.1 | - | 2 | 350 | |||
AY358340.1 | - | 2 | 350 | |||
BC009371.2 | - | 2 | 172 | 4122648 | ||
BE272517.1 | - | 2 | 420 | |||
BG723094.1 | - | 2 | 531 | |||
CA395814.1 | - | 2 | 439 |