Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALG13 SIRNA Assays ›
Gene Symbol:
ALG13
Gene Name:
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases:
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location:
Chr.X: 111680741 - 111764164 on Build GRCh38
UniGene ID:
Hs.443061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36527
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099922.2
NP_001093392.1
16
2010
NM_001257230.1
NP_001244159.1
16
1906
NM_001257231.1
NP_001244160.1
16
2024
NM_001257234.1
NP_001244163.1
16
1979
NM_001257237.1
NP_001244166.1
16
2083
NM_001324292.1
NP_001311221.1
16
2010
NM_001324293.1
NP_001311222.1
14
1909
XM_006724693.2
XP_006724756.1
16
2383
XM_006724697.3
XP_006724760.2
16
2283
XM_006724698.3
XP_006724761.2
16
2408
XM_011531028.2
XP_011529330.1
16
2384
XM_011531030.1
XP_011529332.1
16
2382
XM_011531031.1
XP_011529333.1
16
2083
XM_011531032.1
XP_011529334.1
16
1889
XM_011531033.2
XP_011529335.1
16
1929
XM_011531034.2
XP_011529336.1
14
1623
XM_011531037.1
XP_011529339.1
6
640
XM_017029846.1
XP_016885335.1
16
2385
XM_017029847.1
XP_016885336.1
16
2384
XM_017029848.1
XP_016885337.1
16
2382
XM_017029849.1
XP_016885338.1
16
2383
XM_017029850.1
XP_016885339.1
16
2382
XM_017029851.1
XP_016885340.1
17
2291
XM_017029852.1
XP_016885341.1
9
956
XM_017029853.1
XP_016885342.1
12
1198
XM_017029854.1
XP_016885343.1
3
720
XM_017029855.1
XP_016885344.1
9
1648
GenBank mRNA
AK026671.1
-
1
139
AK223154.1
-
1
139
AK300394.1
-
16
1848
AK302729.1
-
16
1951
AK302890.1
-
16
1927
AK304712.1
-
16
1900
AK316306.1
-
16
1952
AK316522.1
-
16
2027
BC117377.1
-
1
137
40125928
BC117379.1
-
1
137
40125930
BP344481.1
-
2
219
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
transferase
Gene Ontology Categories:
Function(s)
Process(es)
dolichol-linked oligosaccharide biosynthetic process
proteolysis
N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase activity
thiol-dependent ubiquitinyl hydrolase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System