Search
Search

BTNL8| Gene Symbol: BTNL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040462.2 | NP_001035552.1 | 8 | 1384 | ||
| NM_001159707.1 | NP_001153179.1 | 7 | 1036 | |||
| NM_001159708.1 | NP_001153180.1 | 7 | 1493 | |||
| NM_001159709.1 | NP_001153181.1 | 8 | 1087 | |||
| NM_001159710.1 | NP_001153182.1 | 7 | 887 | |||
| NM_024850.2 | NP_079126.1 | 8 | 1514 | |||
| XM_011534649.1 | XP_011532951.1 | 7 | 1270 | |||
| XM_011534650.2 | XP_011532952.1 | 8 | 1189 | |||
| XM_011534651.2 | XP_011532953.1 | 7 | 2203 | |||
| XM_011534652.2 | XP_011532954.1 | 7 | 2179 | |||
| XM_011534653.2 | XP_011532955.1 | 6 | 740 | |||
| GenBank mRNA | AK025111.1 | - | 8 | 1432 | ||
| AK225213.1 | - | 8 | 1432 | |||
| AK296597.1 | - | 8 | 1042 | |||
| AK298687.1 | - | 7 | 887 | |||
| AY358523.1 | - | 8 | 1332 | |||
| BC119696.1 | - | 7 | 839 | 40116820 | ||