Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DHDDS SIRNA Assays ›
Gene Symbol:
DHDDS
Gene Name:
dehydrodolichyl diphosphate synthase subunit
Gene Aliases:
CIT, CPT, DS, HDS, RP59
Chromosome Location:
Chr.1: 26432282 - 26471306 on Build GRCh38
UniGene ID:
Hs.369385
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36696
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DHDDS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243564.1
NP_001230493.1
3
247
NM_001243565.1
NP_001230494.1
3
247
NM_001319959.1
NP_001306888.1
3
247
NM_024887.3
NP_079163.2
3
247
NM_205861.2
NP_995583.1
3
247
XM_006710912.2
XP_006710975.1
3
306
XM_006710913.2
XP_006710976.1
3
185
XM_006710914.2
XP_006710977.1
3
182
XM_006710915.2
XP_006710978.1
3
219
XM_006710916.2
XP_006710979.1
3
273
XM_006710917.2
XP_006710980.1
3
226
XM_006710918.2
XP_006710981.1
3
182
XM_011542183.2
XP_011540485.1
3
282
XM_011542184.2
XP_011540486.1
3
303
XM_011542185.2
XP_011540487.1
3
303
XM_011542186.2
XP_011540488.1
3
303
XM_017002379.1
XP_016857868.1
3
185
XM_017002380.1
XP_016857869.1
3
282
XM_017002381.1
XP_016857870.1
3
273
XM_017002382.1
XP_016857871.1
3
182
XM_017002383.1
XP_016857872.1
3
273
XM_017002384.1
XP_016857873.1
3
226
GenBank mRNA
AB090852.1
-
2
108
AK023164.1
-
3
177
AK297134.1
-
2
167
AK316485.1
-
3
190
BC003643.2
-
2
152
3532466
BC004117.2
-
3
189
3843062
BX648507.1
-
3
249
DA756178.1
-
3
227
GU727641.1
-
2
152
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
dolichyl diphosphate biosynthetic process
polyprenyltransferase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0