Search
Search

DNAJC22| Gene Symbol: DNAJC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304944.1 | NP_001291873.1 | 3 | 1773 | ||
| NM_024902.3 | NP_079178.2 | 2 | 1311 | |||
| XM_005269155.2 | XP_005269212.1 | 2 | 822 | |||
| XM_005269156.2 | XP_005269213.1 | 2 | 822 | |||
| XM_005269157.2 | XP_005269214.1 | 2 | 822 | |||
| GenBank mRNA | AK055747.1 | - | 3 | 1773 | ||
| BC033236.1 | - | 2 | 970 | 5427606 | ||