Search
Search

SCD5| Gene Symbol: SCD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037582.2 | NP_001032671.2 | 3 | 801 | ||
| NM_024906.2 | NP_079182.2 | 3 | 801 | |||
| GenBank mRNA | AF389338.1 | - | 3 | 716 | ||
| AK024685.1 | - | 3 | 716 | |||
| BC137429.1 | - | 3 | 705 | 9021427 | ||
| BC137432.1 | - | 3 | 705 | 9021430 | ||
| DR763368.1 | - | 2 | 214 | |||