Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEMA6D SIRNA Assays ›
Gene Symbol:
SEMA6D
Gene Name:
semaphorin 6D
Gene Aliases:
-
Chromosome Location:
Chr.15: 47184101 - 47774223 on Build GRCh38
UniGene ID:
Hs.511265
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36855
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEMA6D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198999.1
NP_001185928.1
5, 6
561
NM_020858.1
NP_065909.1
2, 3
549
NM_024966.2
NP_079242.2
2, 3
549
NM_153616.1
NP_705869.1
2, 3
549
NM_153617.1
NP_705870.1
2, 3
549
NM_153618.1
NP_705871.1
2, 3
549
NM_153619.1
NP_705872.1
2, 3
549
XM_005254685.3
XP_005254742.1
2, 3
511
XM_005254686.2
XP_005254743.1
2, 3
455
XM_005254687.2
XP_005254744.1
2, 3
455
XM_005254689.3
XP_005254746.1
2, 3
511
XM_011522075.2
XP_011520377.1
5, 6
666
XM_011522076.2
XP_011520378.1
2, 3
455
XM_011522077.2
XP_011520379.1
2, 3
455
XM_011522078.2
XP_011520380.1
2, 3
455
XM_011522079.2
XP_011520381.1
2, 3
455
XM_011522080.2
XP_011520382.1
2, 3
455
XM_011522081.2
XP_011520383.1
2, 3
455
XM_017022617.1
XP_016878106.1
5, 6
479
XM_017022618.1
XP_016878107.1
2, 3
511
XM_017022619.1
XP_016878108.1
2, 3
511
XM_017022620.1
XP_016878109.1
5, 6
666
XM_017022621.1
XP_016878110.1
5, 6
666
GenBank mRNA
AB040912.2
-
5, 6
561
AF389426.1
-
2, 3
549
AF389427.1
-
2, 3
549
AF389428.1
-
2, 3
549
AF389429.1
-
2, 3
549
AF389430.1
-
2, 3
549
AK290032.1
-
2, 3
470
BC150253.1
-
5, 6
561
9007179
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_199238, NM_199240, NM_172537, NM_199241, NM_199239, and others.
Panther Classification:
Molecular Function -
membrane-bound signaling molecule
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
neural crest cell migration
positive regulation of smooth muscle cell migration
negative regulation of smooth muscle cell migration
ventricular system development
positive regulation of cell migration
negative regulation of axon extension involved in axon guidance
negative chemotaxis
semaphorin-plexin signaling pathway
semaphorin receptor binding
chemorepellent activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0