Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MAP3K19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018044.2 | NP_001018054.1 | 7 | 3266 | ||
NM_001018046.2 | NP_001018056.1 | 8 | 1151 | |||
NM_001018047.2 | NP_001018057.1 | 8 | 1007 | |||
NM_001282883.1 | NP_001269812.1 | 7 | 1001 | |||
NM_001321177.1 | NP_001308106.1 | 10 | 3897 | |||
NM_025052.4 | NP_079328.3 | 9 | 3605 | |||
XM_011511891.2 | XP_011510193.1 | 9 | 3745 | |||
XM_011511892.1 | XP_011510194.1 | 12 | 4313 | |||
XM_011511894.2 | XP_011510196.1 | 9 | 3720 | |||
XM_011511896.2 | XP_011510198.1 | 9 | 3586 | |||
XM_011511897.2 | XP_011510199.1 | 9 | 3666 | |||
XM_017005003.1 | XP_016860492.1 | 12 | 4280 | |||
XM_017005004.1 | XP_016860493.1 | 11 | 4165 | |||
XM_017005005.1 | XP_016860494.1 | 10 | 3966 | |||
XM_017005006.1 | XP_016860495.1 | 10 | 3328 | |||
XM_017005007.1 | XP_016860496.1 | 9 | 3713 | |||
GenBank mRNA | AK026727.1 | - | 1 | 486 | ||
AK122935.1 | - | 10 | 3897 | |||
AK131216.1 | - | 3 | 1744 | |||
AK225579.1 | - | 1 | 486 | |||
AY574901.1 | - | 9 | 3605 | |||
AY574902.1 | - | 7 | 3266 | |||
AY574904.1 | - | 8 | 1151 | |||
AY574905.1 | - | 8 | 1007 | |||
BC034417.1 | - | 2 | 652 | |||
BC137276.1 | - | 9 | 3604 | 9021273 | ||
BC137277.1 | - | 9 | 3604 | 9021274 | ||
BC144555.1 | - | 7 | 1000 |