Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZC3H12A SIRNA Assays ›
          Gene Symbol:
          ZC3H12A
          Gene Name:
          zinc finger CCCH-type containing 12A
          Gene Aliases:
          MCPIP, MCPIP1, dJ423B22.1
          Chromosome Location:
          Chr.1: 37474518 - 37484377 on Build GRCh38
          UniGene ID:
          Hs.656294
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s36969

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZC3H12A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001323550.1 NP_001310479.1 5, 6 1070
          NM_001323551.1 NP_001310480.1 5, 6 1157
          NM_025079.2 NP_079355.2 5, 6 1073
          XM_011542198.2 XP_011540500.1 5, 6 1153
          GenBank mRNA AK026884.1 - 5, 6 1039
          AY920403.1 - 4, 5 923
          BC005001.1 - 5, 6 997 2958199
          CR457364.1 - 4, 5 923

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          endoribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, endonucleolytic cleavage-dependent decay
          angiogenesis
          negative regulation of protein phosphorylation
          positive regulation of defense response to virus by host
          transcription, DNA-templated
          apoptotic process
          inflammatory response
          cellular response to DNA damage stimulus
          nervous system development
          regulation of gene expression
          positive regulation of autophagy
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of muscle cell apoptotic process
          positive regulation of lipid storage
          positive regulation of cell death
          protein deubiquitination
          cell differentiation
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          cellular response to oxidative stress
          cellular response to glucose starvation
          positive regulation of protein import into nucleus
          negative regulation of NF-kappaB import into nucleus
          negative regulation of macrophage activation
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation by host of viral genome replication
          negative regulation of nitric oxide biosynthetic process
          positive regulation of fat cell differentiation
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of interleukin-1 beta secretion
          protein oligomerization
          defense response to virus
          negative regulation of cardiac muscle contraction
          positive regulation of mRNA catabolic process
          3'-UTR-mediated mRNA destabilization
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          RNA phosphodiester bond hydrolysis
          RNA phosphodiester bond hydrolysis, endonucleolytic
          RNA phosphodiester bond hydrolysis, exonucleolytic
          cellular response to virus
          negative regulation of cytokine production involved in inflammatory response
          positive regulation of execution phase of apoptosis
          negative regulation of interleukin-6 secretion
          positive regulation of p38MAPK cascade
          negative regulation of interferon-gamma secretion
          positive regulation of protein deubiquitination
          negative regulation of production of miRNAs involved in gene silencing by miRNA
          cellular response to sodium arsenite
          negative regulation of tumor necrosis factor secretion
          cellular response to phorbol 13-acetate 12-myristate
          cellular response to ionomycin
          cellular response to chemokine
          positive regulation of reactive oxygen species metabolic process
          positive regulation of miRNA catabolic process
          DNA binding
          chromatin binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          endoribonuclease activity
          exoribonuclease activity
          ribonuclease activity
          thiol-dependent ubiquitin-specific protease activity
          protein binding
          miRNA binding
          RNA stem-loop binding
          mRNA 3'-UTR AU-rich region binding
          ribosome binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0