Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZC3H12A SIRNA Assays ›
          Gene Symbol:
          ZC3H12A
          Gene Name:
          zinc finger CCCH-type containing 12A
          Gene Aliases:
          MCPIP, MCPIP1, dJ423B22.1
          Chromosome Location:
          Chr.1: 37474518 - 37484377 on Build GRCh38
          UniGene ID:
          Hs.656294
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s36971

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZC3H12A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001323550.1 NP_001310479.1 2 330
          NM_001323551.1 NP_001310480.1 2 333
          NM_025079.2 NP_079355.2 2 333
          XM_011542198.2 XP_011540500.1 2 333
          GenBank mRNA AK026884.1 - 2 299
          AY920403.1 - 1 183
          BC005001.1 - 2 257 2958199
          CR457364.1 - 1 183
          FJ695517.1 - 2 271

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          endoribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, endonucleolytic cleavage-dependent decay
          angiogenesis
          negative regulation of protein phosphorylation
          positive regulation of defense response to virus by host
          transcription, DNA-templated
          apoptotic process
          inflammatory response
          cellular response to DNA damage stimulus
          nervous system development
          regulation of gene expression
          positive regulation of autophagy
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of muscle cell apoptotic process
          positive regulation of lipid storage
          positive regulation of cell death
          protein deubiquitination
          cell differentiation
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          cellular response to oxidative stress
          cellular response to glucose starvation
          positive regulation of protein import into nucleus
          negative regulation of NF-kappaB import into nucleus
          negative regulation of macrophage activation
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation by host of viral genome replication
          negative regulation of nitric oxide biosynthetic process
          positive regulation of fat cell differentiation
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of interleukin-1 beta secretion
          protein oligomerization
          defense response to virus
          negative regulation of cardiac muscle contraction
          positive regulation of mRNA catabolic process
          3'-UTR-mediated mRNA destabilization
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          RNA phosphodiester bond hydrolysis
          RNA phosphodiester bond hydrolysis, endonucleolytic
          RNA phosphodiester bond hydrolysis, exonucleolytic
          cellular response to virus
          negative regulation of cytokine production involved in inflammatory response
          positive regulation of execution phase of apoptosis
          negative regulation of interleukin-6 secretion
          positive regulation of p38MAPK cascade
          negative regulation of interferon-gamma secretion
          positive regulation of protein deubiquitination
          negative regulation of production of miRNAs involved in gene silencing by miRNA
          cellular response to sodium arsenite
          negative regulation of tumor necrosis factor secretion
          cellular response to phorbol 13-acetate 12-myristate
          cellular response to ionomycin
          cellular response to chemokine
          positive regulation of reactive oxygen species metabolic process
          positive regulation of miRNA catabolic process
          DNA binding
          chromatin binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          endoribonuclease activity
          exoribonuclease activity
          ribonuclease activity
          thiol-dependent ubiquitin-specific protease activity
          protein binding
          miRNA binding
          RNA stem-loop binding
          mRNA 3'-UTR AU-rich region binding
          ribosome binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0