Search
Search

ULBP1| Gene Symbol: ULBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025218.3 | NP_079494.1 | 2 | 371 | ||
| NR_133659.1 | 2 | 1719 | ||||
| XM_017011322.1 | XP_016866811.1 | 2 | 371 | |||
| GenBank mRNA | AB052907.1 | - | 2 | 295 | ||
| AF304377.1 | - | 2 | 295 | |||
| AF346416.1 | - | 2 | 295 | |||
| AF346595.1 | - | 2 | 265 | |||
| AF425265.1 | - | 2 | 282 | |||
| AK292519.1 | - | 2 | 371 | |||
| AK316582.1 | - | 2 | 360 | |||
| BC035416.1 | - | 2 | 338 | 4822311 | ||
| KT591165.1 | - | 2 | 1686 | |||