Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CMIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030629.2 | NP_085132.1 | 7 | 656 | ||
NM_198390.2 | NP_938204.2 | 7 | 882 | |||
XM_005256179.4 | XP_005256236.4 | 7 | 1837 | |||
XM_005256181.2 | XP_005256238.1 | 6 | 613 | |||
XM_005256182.1 | XP_005256239.1 | 5 | 370 | |||
XM_011523352.1 | XP_011521654.1 | 7 | 842 | |||
XM_011523353.1 | XP_011521655.1 | 5 | 750 | |||
XM_017023733.1 | XP_016879222.1 | 5 | 5227 | |||
GenBank mRNA | AB051481.1 | - | 7 | 763 | ||
AK096598.1 | - | 7 | 843 | |||
AK304418.1 | - | 6 | 618 | |||
AY172689.1 | - | 7 | 656 | |||
BC038113.1 | - | 5 | 579 | 4818904 |