Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CTSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317331.1 | NP_001304260.1 | 5 | 715 | ||
NM_001910.3 | NP_001901.1 | 6 | 872 | |||
NM_148964.2 | NP_683865.1 | 6 | 872 | |||
XM_011509244.1 | XP_011507546.1 | 6 | 877 | |||
XM_011509245.1 | XP_011507547.1 | 6 | 877 | |||
GenBank mRNA | AJ250716.1 | - | 6 | 858 | ||
AJ250717.1 | - | 6 | 858 | |||
AK292057.1 | - | 6 | 802 | |||
AK298069.1 | - | 5 | 568 | |||
BC042537.1 | - | 6 | 839 | 5174814 | ||
J05036.1 | - | 6 | 803 |