Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LBH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030915.3 | NP_112177.2 | 3 | 977 | ||
XM_017005059.1 | XP_016860548.1 | 3 | 1135 | |||
GenBank mRNA | AK023425.1 | - | 1 | 44 | ||
AK027232.1 | - | 3 | 769 | |||
AK223541.1 | - | 3 | 913 | |||
AL136703.1 | - | 3 | 982 | |||
BC109376.1 | - | 3 | 981 | 6495944 | ||
BC126434.1 | - | 3 | 950 | 8992150 | ||
BC130415.1 | - | 3 | 949 | 40146446 | ||
EF025586.1 | - | 3 | 955 |