Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNF146 SIRNA Assays ›
Gene Symbol:
RNF146
Gene Name:
ring finger protein 146
Gene Aliases:
-
Chromosome Location:
Chr.6: 127266340 - 127289893 on Build GRCh38
UniGene ID:
Hs.267120
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s37823
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNF146
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242844.1
NP_001229773.1
5
1143
NM_001242845.1
NP_001229774.1
5
992
NM_001242846.1
NP_001229775.1
5
905
NM_001242847.1
NP_001229776.1
4
1034
NM_001242848.1
NP_001229777.1
4
764
NM_001242849.1
NP_001229778.1
3
906
NM_001242850.1
NP_001229779.1
3
636
NM_001242851.1
NP_001229780.1
3
576
NM_001242852.1
NP_001229781.1
5
1123
NM_030963.3
NP_112225.2
5
873
XM_006715571.3
XP_006715634.1
5
1573
XM_011536161.2
XP_011534463.1
6
1460
XM_011536162.2
XP_011534464.1
6
1512
XM_011536163.2
XP_011534465.1
6
1492
XM_011536164.2
XP_011534466.1
5
1351
XM_017011336.1
XP_016866825.1
3
1253
XM_017011337.1
XP_016866826.1
5
4494
XM_017011338.1
XP_016866827.1
7
1601
XM_017011339.1
XP_016866828.1
6
1630
XM_017011340.1
XP_016866829.1
6
1682
XM_017011341.1
XP_016866830.1
4
1381
XM_017011342.1
XP_016866831.1
5
1522
XM_017011343.1
XP_016866832.1
6
1631
GenBank mRNA
AJ315122.1
-
1
438
AJ420549.1
-
2
508
AK027436.1
-
2
577
AK027558.1
-
3
625
AK027776.1
-
1
448
AL136829.1
-
5
836
BC008235.1
-
2
525
3868592
BI771613.1
-
2
575
CN388414.1
-
3
716
CR533514.1
-
1
437
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
Wnt signaling pathway
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
protein autoubiquitination
protein K48-linked ubiquitination
positive regulation of canonical Wnt signaling pathway
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
poly-ADP-D-ribose binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0