Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127711.2 | NP_001121183.1 | 4 | 314 | ||
NM_001286484.1 | NP_001273413.1 | 4 | 339 | |||
NM_001286488.1 | NP_001273417.1 | 5 | 416 | |||
NM_001286489.1 | NP_001273418.1 | 4 | 314 | |||
NM_030969.4 | NP_112231.3 | 5 | 416 | |||
NR_104454.1 | 3 | 237 | ||||
GenBank mRNA | AK294152.1 | - | 5 | 370 | ||
BC001033.1 | - | 5 | 358 | 3139565 | ||
BC013913.1 | - | 5 | 378 | 3685635 | ||
BC071660.1 | - | 5 | 360 | 4473732 | ||
BG288654.1 | - | 4 | 267 | |||
BG432075.1 | - | 5 | 345 | |||
BP249229.1 | - | 4 | 268 | |||
BQ056988.1 | - | 4 | 267 | |||
BU954633.1 | - | 3 | 179 |