Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: INHBE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031479.3 | NP_113667.1 | 2 | 1206 | ||
GenBank mRNA | AF412024.1 | - | 2 | 1190 | ||
AK075285.1 | - | 2 | 1201 | |||
AK308647.1 | - | 2 | 901 | |||
BC005161.2 | - | 2 | 1198 | 3529092 |