Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RSPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031924.4 | NP_114130.3 | 7 | 1508 | ||
XM_005267153.4 | XP_005267210.1 | 5 | 1486 | |||
XM_017011347.1 | XP_016866836.1 | 6 | 829 | |||
GenBank mRNA | AF353618.1 | - | 7 | 1107 | ||
AK057655.1 | - | 5 | 835 | |||
AK057931.1 | - | 7 | 1529 | |||
BC050604.1 | - | 7 | 1508 | 6161669 |