Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SPATA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031955.5 | NP_114161.3 | 7, 8 | 1408 | ||
XM_006713778.2 | XP_006713841.1 | 7, 8 | 1382 | |||
XM_017007308.1 | XP_016862797.1 | 7, 8 | 1382 | |||
GenBank mRNA | AF345909.2 | - | 7, 8 | 1354 | ||
AF404758.1 | - | 7, 8 | 1354 | |||
AK093041.1 | - | 7, 8 | 1356 | |||
BC034496.2 | - | 7, 8 | 1310 | 4825905 |