Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031965.2 | NP_114171.2 | 1 | 2435 | ||
GenBank mRNA | AF289865.1 | - | 1 | 2431 | ||
AK056691.1 | - | 2 | 1321 | |||
AK292715.1 | - | 1 | 2421 | |||
BC047457.1 | - | 1 | 2416 | 5298590 |