Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OBSCN SIRNA Assays ›
          Gene Symbol:
          OBSCN
          Gene Name:
          obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
          Gene Aliases:
          ARHGEF30, UNC89
          Chromosome Location:
          Chr.1: 228208013 - 228381431 on Build GRCh38
          UniGene ID:
          Hs.650039
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s38403

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: OBSCN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098623.2 NP_001092093.2 56, 57 15725
          NM_001271223.2 NP_001258152.2 67, 68 18596
          NM_052843.3 NP_443075.3 56, 57 15725
          XM_005273287.4 XP_005273344.1 68, 69 18976
          XM_005273291.4 XP_005273348.1 68, 69 18940
          XM_005273298.4 XP_005273355.1 67, 68 18667
          XM_005273307.4 XP_005273364.1 63, 64 17854
          XM_006711822.3 XP_006711885.1 66, 67 18400
          XM_006711823.3 XP_006711886.1 66, 67 18400
          XM_006711827.3 XP_006711890.1 60, 61 16815
          XM_006711829.3 XP_006711892.1 59, 60 16539
          XM_011544297.2 XP_011542599.1 60, 61 16816
          XM_011544299.2 XP_011542601.1 59, 60 16540
          XM_017002443.1 XP_016857932.1 68, 69 19021
          XM_017002444.1 XP_016857933.1 68, 69 18984
          XM_017002445.1 XP_016857934.1 68, 69 19021
          XM_017002446.1 XP_016857935.1 68, 69 19021
          XM_017002447.1 XP_016857936.1 68, 69 19018
          XM_017002448.1 XP_016857937.1 68, 69 18871
          XM_017002449.1 XP_016857938.1 67, 68 18758
          XM_017002450.1 XP_016857939.1 67, 68 18758
          XM_017002451.1 XP_016857940.1 67, 68 18758
          XM_017002452.1 XP_016857941.1 67, 68 18758
          XM_017002453.1 XP_016857942.1 67, 68 18745
          XM_017002454.1 XP_016857943.1 67, 68 18745
          XM_017002455.1 XP_016857944.1 67, 68 18745
          XM_017002456.1 XP_016857945.1 67, 68 18745
          XM_017002457.1 XP_016857946.1 67, 68 18745
          XM_017002458.1 XP_016857947.1 66, 67 18494
          XM_017002459.1 XP_016857948.1 66, 67 18494
          XM_017002460.1 XP_016857949.1 66, 67 18494
          XM_017002461.1 XP_016857950.1 66, 67 18481
          XM_017002462.1 XP_016857951.1 66, 67 18469
          XM_017002463.1 XP_016857952.1 66, 67 18448
          XM_017002464.1 XP_016857953.1 66, 67 18397
          XM_017002465.1 XP_016857954.1 65, 66 18230
          XM_017002466.1 XP_016857955.1 65, 66 18230
          XM_017002467.1 XP_016857956.1 65, 66 18230
          XM_017002468.1 XP_016857957.1 64, 65 17966
          XM_017002469.1 XP_016857958.1 62, 63 17438
          XM_017002470.1 XP_016857959.1 57, 58 16119
          XM_017002471.1 XP_016857960.1 68, 69 19020
          XM_017002472.1 XP_016857961.1 68, 69 19020
          XM_017002473.1 XP_016857962.1 68, 69 19020
          GenBank mRNA AJ002535.1 - 57, 58 15722
          AL832357.1 - 1, 2 141

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          multicellular organism development
          regulation of Rho protein signal transduction
          protein localization to M-band
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          sarcomere organization
          regulation of small GTPase mediated signal transduction
          mitophagy in response to mitochondrial depolarization
          protein serine/threonine kinase activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          protein binding
          calmodulin binding
          ATP binding
          structural constituent of muscle
          ankyrin binding
          titin binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline