Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MELK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256685.1 | NP_001243614.1 | 5 | 522 | ||
NM_001256689.1 | NP_001243618.1 | 4 | 436 | |||
NM_001256690.1 | NP_001243619.1 | 3 | 319 | |||
NM_001256693.1 | NP_001243622.1 | 3 | 340 | |||
NM_014791.3 | NP_055606.1 | 5 | 522 | |||
NR_046337.1 | 4 | 436 | ||||
XM_011518076.2 | XP_011516378.1 | 5 | 701 | |||
XM_011518077.1 | XP_011516379.1 | 6 | 597 | |||
XM_011518078.2 | XP_011516380.1 | 6 | 784 | |||
XM_011518079.1 | XP_011516381.1 | 5 | 474 | |||
XM_011518080.1 | XP_011516382.1 | 5 | 489 | |||
XM_011518081.2 | XP_011516383.1 | 5 | 507 | |||
XM_011518082.2 | XP_011516384.1 | 4 | 615 | |||
XM_011518083.2 | XP_011516385.1 | 5 | 698 | |||
XM_011518084.2 | XP_011516386.1 | 4 | 310 | |||
XM_011518085.1 | XP_011516387.1 | 3 | 306 | |||
XM_011518086.2 | XP_011516388.1 | 5 | 489 | |||
GenBank mRNA | AB183427.1 | - | 4 | 387 | ||
AB183428.1 | - | 3 | 272 | |||
AK222811.1 | - | 5 | 521 | |||
AK293447.1 | - | 3 | 276 | |||
AK296820.1 | - | 4 | 435 | |||
AK299164.1 | - | 5 | 474 | |||
AK300821.1 | - | 4 | 390 | |||
AK301131.1 | - | 3 | 272 | |||
AK309470.1 | - | 3 | 297 | |||
BC014039.2 | - | 5 | 476 | 4547136 | ||
BP365611.1 | - | 5 | 524 | |||
D79997.1 | - | 5 | 508 |