Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032310.4 | NP_115686.3 | 3, 4 | 534 | ||
NR_134466.1 | 3, 4 | 545 | ||||
NR_134467.1 | 4, 5 | 636 | ||||
GenBank mRNA | BC004500.2 | - | 4, 5 | 403 | 3833318 | |
BC091476.1 | - | 3, 4 | 388 | |||
BX354102.2 | - | 4, 5 | 516 | |||
DB550397.1 | - | 1, 2 | 30 |