Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM2 SIRNA Assays ›
Gene Symbol:
ABLIM2
Gene Name:
actin binding LIM protein family member 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 7965310 - 8158832 on Build GRCh38
UniGene ID:
Hs.233404
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s39023
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130083.1
NP_001123555.1
3
405
NM_001130084.1
NP_001123556.1
3
405
NM_001130085.1
NP_001123557.1
3
405
NM_001130086.1
NP_001123558.1
3
405
NM_001130087.1
NP_001123559.1
3
405
NM_001130088.1
NP_001123560.1
3
405
NM_032432.4
NP_115808.3
3
405
XM_005248014.4
XP_005248071.1
3
376
XM_005248017.4
XP_005248074.1
3
376
XM_005248018.4
XP_005248075.1
3
375
XM_005248019.4
XP_005248076.1
3
375
XM_005248020.4
XP_005248077.1
3
375
XM_005248021.4
XP_005248078.1
3
374
XM_005248022.4
XP_005248079.1
3
375
XM_005248023.4
XP_005248080.1
3
375
XM_005248024.4
XP_005248081.1
3
374
XM_005248027.4
XP_005248084.1
3
374
XM_005248028.4
XP_005248085.1
3
371
XM_005248029.4
XP_005248086.1
3
373
XM_005248030.4
XP_005248087.1
3
373
XM_005248031.4
XP_005248088.1
3
375
XM_006713923.3
XP_006713986.1
3
376
XM_006713924.3
XP_006713987.1
3
375
XM_006713925.3
XP_006713988.1
3
376
XM_006713927.3
XP_006713990.1
3
374
XM_011513586.2
XP_011511888.1
3
380
XM_017008706.1
XP_016864195.1
3
377
XM_017008707.1
XP_016864196.1
3
386
XM_017008708.1
XP_016864197.1
3
375
XM_017008709.1
XP_016864198.1
3
376
XM_017008710.1
XP_016864199.1
3
375
XM_017008711.1
XP_016864200.1
3
375
XM_017008712.1
XP_016864201.1
3
375
XM_017008713.1
XP_016864202.1
3
374
XM_017008714.1
XP_016864203.1
3
386
XM_017008715.1
XP_016864204.1
3
375
XM_017008716.1
XP_016864205.1
3
374
XM_017008717.1
XP_016864206.1
3
374
XM_017008718.1
XP_016864207.1
3
386
XM_017008719.1
XP_016864208.1
3
374
XM_017008720.1
XP_016864209.1
3
373
XM_017008721.1
XP_016864210.1
3
373
XM_017008722.1
XP_016864211.1
3
374
XM_017008723.1
XP_016864212.1
3
386
XM_017008724.1
XP_016864213.1
3
386
XM_017008725.1
XP_016864214.1
3
386
GenBank mRNA
AB058711.1
-
2
164
AJ748600.1
-
3
327
AJ748601.1
-
3
327
AK094754.1
-
3
384
AK301060.1
-
3
368
BC067214.1
-
3
417
BC122567.1
-
3
356
40085570
DQ413176.1
-
3
282
DQ413177.1
-
2
262
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
actin cytoskeleton organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-4p4qf:80/100.66.72.111:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0