Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL17RC SIRNA Assays ›
          Gene Symbol:
          IL17RC
          Gene Name:
          interleukin 17 receptor C
          Gene Aliases:
          CANDF9, IL17-RL, IL17RL
          Chromosome Location:
          Chr.3: 9917074 - 9933627 on Build GRCh38
          UniGene ID:
          Hs.129959
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s39470

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL17RC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001203263.1 NP_001190192.1 8 882
          NM_001203264.1 NP_001190193.1 8 882
          NM_001203265.1 NP_001190194.1 7 837
          NM_032732.5 NP_116121.2 7 837
          NM_153460.3 NP_703190.1 8 882
          NM_153461.3 NP_703191.1 8 1071
          NR_037807.1 7 837
          XM_017007326.1 XP_016862815.1 8 1071
          XM_017007327.1 XP_016862816.1 8 1071
          XM_017007328.1 XP_016862817.1 8 1071
          XM_017007329.1 XP_016862818.1 8 1071
          XM_017007330.1 XP_016862819.1 8 1071
          XM_017007331.1 XP_016862820.1 7 1026
          XM_017007332.1 XP_016862821.1 7 1026
          XM_017007333.1 XP_016862822.1 8 1071
          XM_017007334.1 XP_016862823.1 7 996
          XM_017007335.1 XP_016862824.1 7 1026
          XM_017007336.1 XP_016862825.1 8 1071
          XM_017007337.1 XP_016862826.1 6 951
          XM_017007338.1 XP_016862827.1 7 1026
          XM_017007339.1 XP_016862828.1 7 1026
          XM_017007340.1 XP_016862829.1 8 859
          XM_017007341.1 XP_016862830.1 7 814
          XM_017007342.1 XP_016862831.1 7 783
          XM_017007343.1 XP_016862832.1 7 783
          XM_017007344.1 XP_016862833.1 8 859
          XM_017007345.1 XP_016862834.1 7 814
          XM_017007346.1 XP_016862835.1 7 814
          XM_017007347.1 XP_016862836.1 6 738
          XM_017007348.1 XP_016862837.1 7 814
          XM_017007349.1 XP_016862838.1 6 738
          GenBank mRNA AF458065.1 - 8 837
          AK075534.1 - 7 802
          AK092907.1 - 8 1072
          AK096824.1 - 7 862
          AK225650.1 - 8 827
          AK290670.1 - 7 874
          AK294616.1 - 6 680
          AK295377.1 - 6 739
          AK303171.1 - 6 648
          AK308241.1 - 7 767
          AK315024.1 - 7 814
          AK315053.1 - 7 782
          AY358840.1 - 7 799
          AY359098.1 - 7 827
          EF676032.1 - 7 748
          EF676033.1 - 8 640
          EF676034.1 - 8 651

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine-mediated signaling pathway
          defense response to fungus
          granulocyte chemotaxis
          positive regulation of cytokine production involved in inflammatory response
          positive regulation of interleukin-6 secretion
          receptor binding
          interleukin-17 receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-d655g:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0