Search
Search

RPUSD4| Gene Symbol: RPUSD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144827.1 | NP_001138299.1 | 3 | 576 | ||
| NM_032795.2 | NP_116184.2 | 3 | 576 | |||
| XM_011543039.2 | XP_011541341.1 | 3 | 546 | |||
| GenBank mRNA | AK027400.1 | - | 3 | 530 | ||
| AK225320.1 | - | 3 | 536 | |||
| AK308059.1 | - | 3 | 529 | |||
| BC014131.2 | - | 3 | 537 | 4621434 | ||
| DB051034.1 | - | 3 | 578 | |||