Search
Search

POMGNT2| Gene Symbol: POMGNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032806.5 | NP_116195.2 | 2 | 2042 | ||
| XM_005265515.2 | XP_005265572.1 | 3 | 2173 | |||
| XM_011534163.2 | XP_011532465.1 | 3 | 2161 | |||
| XM_011534164.1 | XP_011532466.1 | 2 | 2743 | |||
| XM_017007353.1 | XP_016862842.1 | 4 | 2252 | |||
| GenBank mRNA | AK027472.1 | - | 1 | 1302 | ||
| AK092147.1 | - | 2 | 1830 | |||
| AK124737.1 | - | 1 | 3010 | |||
| BC060861.1 | - | 2 | 2032 | 30335722 | ||