Search
Search

POMGNT2| Gene Symbol: POMGNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032806.5 | NP_116195.2 | 2 | 1695 | ||
| XM_005265515.2 | XP_005265572.1 | 3 | 1826 | |||
| XM_011534163.2 | XP_011532465.1 | 3 | 1814 | |||
| XM_011534164.1 | XP_011532466.1 | 2 | 2396 | |||
| XM_017007353.1 | XP_016862842.1 | 4 | 1905 | |||
| GenBank mRNA | AK027472.1 | - | 1 | 955 | ||
| AK092147.1 | - | 2 | 1483 | |||
| AK124737.1 | - | 1 | 2663 | |||
| BC060861.1 | - | 2 | 1685 | 30335722 | ||