Search
Search

ZNF341| Gene Symbol: ZNF341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282933.1 | NP_001269862.1 | 12 | 2082 | ||
| NM_001282935.1 | NP_001269864.1 | 11 | 1885 | |||
| NM_032819.4 | NP_116208.3 | 12 | 2061 | |||
| NR_104259.1 | 11 | 1985 | ||||
| GenBank mRNA | AK027550.1 | - | 6 | 857 | ||
| AK096755.1 | - | 10 | 1538 | |||
| AK122943.1 | - | 12 | 2163 | |||
| BC094738.1 | - | 11 | 1644 | |||
| BC132873.1 | - | 12 | 1882 | 40146895 | ||
| BC144188.1 | - | 12 | 1882 | 9052708 | ||
| BC146811.1 | - | 12 | 1844 | 8860372 | ||
| BC157823.1 | - | 12 | 1861 | 9057042 | ||