Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DOCK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271999.1 | NP_001258928.1 | 12 | 1478 | ||
NM_001272000.1 | NP_001258929.1 | 12 | 1478 | |||
NM_001272001.1 | NP_001258930.1 | 12 | 1478 | |||
NM_001272002.1 | NP_001258931.1 | 12 | 1478 | |||
NM_033407.3 | NP_212132.2 | 12 | 1478 | |||
XM_005271292.2 | XP_005271349.1 | 12 | 1468 | |||
XM_011542326.2 | XP_011540628.1 | 12 | 1468 | |||
XM_011542327.2 | XP_011540629.1 | 12 | 1468 | |||
XM_011542328.2 | XP_011540630.1 | 12 | 1468 | |||
XM_011542330.2 | XP_011540632.1 | 12 | 1468 | |||
XM_017002639.1 | XP_016858128.1 | 12 | 1468 | |||
XM_017002640.1 | XP_016858129.1 | 12 | 1468 | |||
GenBank mRNA | AK055401.1 | - | 12 | 1408 | ||
AK292640.1 | - | 12 | 1468 | |||
DQ118679.1 | - | 9 | 1050 | |||
DQ118680.1 | - | 9 | 1050 | |||
DQ309763.1 | - | 9 | 1050 | |||
DQ341187.1 | - | 12 | 1385 |