Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322178.1 | NP_001309107.1 | 12 | 2229 | ||
NM_001322179.1 | NP_001309108.1 | 13 | 2379 | |||
NM_001322180.1 | NP_001309109.1 | 12 | 2185 | |||
NM_001322181.1 | NP_001309110.1 | 13 | 2376 | |||
NM_001322187.1 | NP_001309116.1 | 14 | 2444 | |||
NM_001322188.1 | NP_001309117.1 | 13 | 2379 | |||
NM_001322189.1 | NP_001309118.1 | 14 | 2638 | |||
NM_001322190.1 | NP_001309119.1 | 12 | 2235 | |||
NM_001322191.1 | NP_001309120.1 | 12 | 2180 | |||
NM_033401.4 | NP_207837.2 | 13 | 2379 | |||
NM_138994.4 | NP_620481.2 | 12 | 1773 | |||
NR_136210.1 | 13 | 2379 | ||||
NR_136213.1 | 13 | 2420 | ||||
NR_136216.1 | 15 | 2608 | ||||
XM_011523403.2 | XP_011521705.1 | 13 | 1917 | |||
XM_017023805.1 | XP_016879294.1 | 13 | 2108 | |||
GenBank mRNA | AB051550.2 | - | 13 | 2318 | ||
AB100092.1 | - | 12 | 1789 | |||
AB100093.1 | - | 11 | 1714 | |||
AB100094.1 | - | 13 | 1933 | |||
AF463518.1 | - | 12 | 2081 | |||
AK057327.1 | - | 13 | 1903 |