Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DDX11 SIRNA Assays ›
          Gene Symbol:
          DDX11 DDX12P
          Gene Name:
          DEAD/H-box helicase 11
          DEAD/H-box helicase 12, pseudogene
          Gene Aliases:
          CHL1, CHLR1, KRG2, WABS
          CHLR2, DDX12
          Chromosome Location:
          Chr.12: 31073574 - 31104799 on Build GRCh38
          Chromosome Location:
          Chr.12: 9417691 - 9448172 on Build GRCh38
          UniGene ID:
          Hs.443960
          Hs.447869
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s4025

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DDX11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257144.1 NP_001244073.1 22 2345
          NM_001257145.1 NP_001244074.1 22 2272
          NM_004399.2 NP_004390.3 21 2317
          NM_030653.3 NP_085911.2 22 2467
          NM_152438.1 NP_689651.1 22 2467
          XM_005253331.3 XP_005253388.1 22 2637
          XM_005253333.3 XP_005253390.1 22 2637
          XM_006719050.3 XP_006719113.1 22 2637
          XM_011520592.2 XP_011518894.1 22 2638
          XM_011520595.2 XP_011518897.1 21 2487
          XM_017018918.1 XP_016874407.1 23 2820
          XM_017018919.1 XP_016874408.1 24 2883
          XM_017018920.1 XP_016874409.1 23 2821
          XM_017018921.1 XP_016874410.1 23 2820
          XM_017018922.1 XP_016874411.1 23 2613
          XM_017018923.1 XP_016874412.1 23 2506
          XM_017018924.1 XP_016874413.1 23 2539
          XM_017018925.1 XP_016874414.1 23 2611
          XM_017018926.1 XP_016874415.1 23 2680
          XM_017018927.1 XP_016874416.1 22 2671
          XM_017018928.1 XP_016874417.1 23 2820
          XM_017018929.1 XP_016874418.1 23 2821
          XM_017018930.1 XP_016874419.1 23 2819
          XM_017018931.1 XP_016874420.1 22 2433
          XM_017018932.1 XP_016874421.1 22 2326
          XM_017018933.1 XP_016874422.1 23 2907
          XM_017018934.1 XP_016874423.1 22 2433
          XM_017018935.1 XP_016874424.1 19 1843
          XM_017018936.1 XP_016874425.1 19 2838
          XM_017018937.1 XP_016874426.1 19 2843
          XM_017018938.1 XP_016874427.1 22 2553
          XM_017018939.1 XP_016874428.1 21 2544
          GenBank mRNA BC050069.1 - 22 2257 5271949
          BC050522.1 - 22 2296 6471750
          BC111733.1 - 22 2467 40035545
          U33833.1 - 22 2394
          U75967.1 - 22 2357
          U75968.1 - 23 2647
          U75969.1 - 15 1391
          X99583.1 - 21 2278
          Gene Symbol: DDX12P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033399.1 22 2411
          GenBank mRNA BC065494.1 - 12 2182
          U33834.1 - 19 1772

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          sister chromatid cohesion
          viral process
          DNA duplex unwinding
          IRE1-mediated unfolded protein response
          cell cycle
          double-stranded DNA binding
          single-stranded DNA binding
          RNA binding
          ATP-dependent DNA helicase activity
          helicase activity
          protein binding
          ATP binding
          DNA-dependent ATPase activity
          metal ion binding
          4 iron, 4 sulfur cluster binding
          DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0