Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DDX11 SIRNA Assays ›
          Gene Symbol:
          DDX11 DDX12P LOC642846
          Gene Name:
          DEAD/H-box helicase 11
          DEAD/H-box helicase 12, pseudogene
          DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11-like
          Gene Aliases:
          CHL1, CHLR1, KRG2, WABS
          CHLR2, DDX12
          -
          Chromosome Location:
          Chr.12: 31073574 - 31104799 on Build GRCh38
          Chromosome Location:
          Chr.12: 9417691 - 9448172 on Build GRCh38
          Chromosome Location:
          Chr.12: 9283657 - 9314088 on Build GRCh38
          UniGene ID:
          Hs.443960
          Hs.447869
          Hs.447869
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s4026

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DDX11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257144.1 NP_001244073.1 6, 7 816
          NM_001257145.1 NP_001244074.1 6, 7 743
          NM_004399.2 NP_004390.3 6, 7 938
          NM_030653.3 NP_085911.2 6, 7 938
          NM_152438.1 NP_689651.1 6, 7 938
          XM_005253331.3 XP_005253388.1 6, 7 1108
          XM_005253333.3 XP_005253390.1 6, 7 1108
          XM_006719050.3 XP_006719113.1 6, 7 1108
          XM_011520592.2 XP_011518894.1 6, 7 1109
          XM_011520595.2 XP_011518897.1 6, 7 1108
          XM_011520597.2 XP_011518899.1 6, 7 1108
          XM_017018918.1 XP_016874407.1 6, 7 1111
          XM_017018919.1 XP_016874408.1 7, 8 1174
          XM_017018920.1 XP_016874409.1 6, 7 1112
          XM_017018921.1 XP_016874410.1 6, 7 1111
          XM_017018922.1 XP_016874411.1 6, 7 904
          XM_017018923.1 XP_016874412.1 6, 7 797
          XM_017018924.1 XP_016874413.1 6, 7 830
          XM_017018925.1 XP_016874414.1 6, 7 902
          XM_017018926.1 XP_016874415.1 6, 7 1112
          XM_017018927.1 XP_016874416.1 6, 7 1112
          XM_017018928.1 XP_016874417.1 6, 7 1111
          XM_017018929.1 XP_016874418.1 6, 7 1112
          XM_017018930.1 XP_016874419.1 6, 7 1110
          XM_017018931.1 XP_016874420.1 6, 7 904
          XM_017018932.1 XP_016874421.1 6, 7 797
          XM_017018933.1 XP_016874422.1 6, 7 1198
          XM_017018934.1 XP_016874423.1 6, 7 904
          XM_017018935.1 XP_016874424.1 2, 3 134
          XM_017018936.1 XP_016874425.1 2, 3 1129
          XM_017018937.1 XP_016874426.1 2, 3 1134
          XM_017018938.1 XP_016874427.1 6, 7 1110
          XM_017018939.1 XP_016874428.1 6, 7 1110
          GenBank mRNA AK021703.1 - 1, 2 102
          AK297607.1 - 6, 7 923
          AK303139.1 - 6, 7 882
          BC050069.1 - 6, 7 728 5271949
          BC050522.1 - 6, 7 767 6471750
          BC111733.1 - 6, 7 938 40035545
          U33833.1 - 6, 7 865
          U75967.1 - 6, 7 828
          U75968.1 - 7, 8 1095
          X99583.1 - 6, 7 899
          Gene Symbol: DDX12P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA U33834.1 - 3, 4 243
          Gene Symbol: LOC642846
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK310166.1 - 4, 5 485

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          sister chromatid cohesion
          viral process
          DNA duplex unwinding
          IRE1-mediated unfolded protein response
          double-stranded DNA binding
          single-stranded DNA binding
          RNA binding
          ATP-dependent DNA helicase activity
          helicase activity
          protein binding
          ATP binding
          DNA-dependent ATPase activity
          metal ion binding
          4 iron, 4 sulfur cluster binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0