Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138369.2 | NP_612378.1 | 3 | 617 | ||
GenBank mRNA | AK301982.1 | - | 3 | 605 | ||
AY303777.1 | - | 3 | 390 | |||
BC007436.2 | - | 3 | 607 | 2961368 |